Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Members of the 'frizzled' gene family encode 7-transmembrane domain proteins that are receptors for Wnt signaling proteins. The Frizzled 5 protein is believed to be the receptor for the Wnt5A ligand. Frizzled 5 has been reported to be expressed in fetal kidney, fetal and adult liver, fetal lung, and adult pancreas. ESTs have been isolated from bone, liver/spleen, placenta, and prostate libraries. Frizzled 5 was cloned from a retina cDNA library. Receptor for Wnt proteins. Component of the Wnt-Fzd-LRP5-LRP6 complex that triggers beta-catenin signaling through inducing aggregation of receptor-ligand complexes into ribosome-sized signalosomes. The beta-catenin canonical signaling pathway leads to the activation of disheveled proteins, inhibition of GSK-3 kinase, nuclear accumulation of beta-catenin and activation of Wnt target genes. A second signaling pathway involving PKC and calcium fluxes has been seen for some family members, but it is not yet clear if it represents a distinct pathway or if it can be integrated in the canonical pathway, as PKC seems to be required for Wnt-mediated inactivation of GSK-3 kinase. Both pathways seem to involve interactions with G-proteins. May be involved in transduction and intercellular transmission of polarity information during tissue morphogenesis and/or in differentiated tissues. Coreceptor along with RYK of Wnt proteins, such as WNT1.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: frizzled 5, seven transmembrane spanning receptor; frizzled 8, seven transmembrane spanning receptor; frizzled family receptor 5; frizzled family receptor 8; frizzled homolog 8; Frizzled-5; Frizzled-8; Fz-5; Fz-8; FzE5; seven-transmembrane receptor frizzled-5; Wnt receptor
Gene Aliases: 5330434N09Rik; AI427138; C2orf31; Fz-5; FZ-8; Fz5; Fz8; FZD5; FZD8; HFZ5; hFZ8; mFz5
UniProt ID: (Human) Q13467, (Human) Q9H461, (Mouse) Q9EQD0, (Rat) Q8CHL0, (Mouse) Q61091, (Rat) Q498S8
Entrez Gene ID: (Human) 7855, (Human) 8325, (Mouse) 14367, (Rat) 317674, (Mouse) 14370, (Rat) 364754
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support