Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: LEIGRFDPE RGRGAAPCQA VEIPMCRGIG YNLTRMPNLL GHTSQGEAAA ELAEFAPLVQ YGCHSHLRFF LCSLYAPMCT DQVSTPIPAC RPMCEQARLR CAPIMEQFNF GWPDSLDCAR LPTRNDPHAL CMEAPENATA GPAEPHKGLG MLPVAPRPAR PPGDLGPGAG GSGTCENREK FQYVEKSRSC APRCGPGVEV FWSRRDKDFA LVWMAVWSAL CFFSTA (23-247 aa encoded by BC026333)
Members of the 'frizzled' gene family encode 7-transmembrane domain proteins that are receptors for Wnt signaling proteins. The FZD9 gene is located within the Williams syndrome common deletion region of chromosome 7, and heterozygous deletion of the FZD9 gene may contribute to the Williams syndrome phenotype. FZD9 is expressed predominantly in brain, testis, eye, skeletal muscle, and kidney.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CD349; CD349 antigen; Frizzled 3; frizzled 9; frizzled 9, seven transmembrane spanning receptor; frizzled family receptor 9; frizzled homolog 9; Frizzled-9; Frizzled-like protein 9; Frizzled3; Frizzled9; Fz-9; FzE6; hFz9; mFz3; rFz9
Gene Aliases: CD349; FZD3; FZD9; mfz9
UniProt ID: (Human) O00144, (Rat) Q8K4C8, (Mouse) Q9R216
Entrez Gene ID: (Human) 8326, (Rat) 266608, (Mouse) 14371
Molecular Function:
transmembrane signal receptor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support