Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Guanine nucleotide-binding proteins (G proteins) form a large family of signal-transducing molecules. They are found as heterotrimers made up of alpha, beta, and gamma subunits. Members of the G protein family have been characterized most extensively on the basis of the alpha subunit, which binds guanine nucleotide, is capable of hydrolyzing GTP, and interacts with specific receptor and effector molecules. The G protein family includes Gs and Gi, the stimulatory and inhibitory GTP-binding regulators of adenylate cyclase; Go, a protein abundant in brain; and transducin-1 and transducin-2, proteins involved in phototransduction in retinal rods and cones, respectively.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Adenylate cyclase-inhibiting G alpha protein; Gi1 protein alpha subunit; guanine nucleotide binding protein (G protein), alpha inhibiting 1; guanine nucleotide binding protein (G protein), alpha inhibiting activity polypeptide 1; guanine nucleotide binding protein, alpha inhibiting 1; Guanine nucleotide-binding protein G(i) subunit alpha-1; guanine nucleotide-binding protein G(i), alpha-1 subunit
Gene Aliases: AU046200; BPGTPB; Gi; Gialpha1; Gnai-1; GNAI1
UniProt ID: (Human) P63096, (Rat) P10824, (Mouse) B2RSH2
Entrez Gene ID: (Human) 2770, (Rat) 25686, (Mouse) 14677
Molecular Function: heterotrimeric G-protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support