Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Immunogen sequence: MNESKPGDSQ NLACVFCRKH DDCPNKYGEK KTKEKWNLTV HYYCLLMSSG IWQRGKEEEG VYGFLIEDIR KEVNRASKLK CCVCKKNGAS IGCVAPRCKR SYHFPCGLQR ECIFQFTGNF ASFCWDHRPV QIITSNNYRE SLPCTICLEF IEPIPSYNIL RSPCCKNAWF HRDCLQVQAI; Positive Samples: U-87MG, LO2, 293T, OVCAR3, Mouse eye, Mouse brain, Mouse liver, Rat testis; Cellular Location: Cytoplasm, Nucleus, nucleolus
E3 ubiquitin-protein ligase which accepts ubiquitin from an E2 ubiquitin-conjugating enzyme in the form of a thioester and then directly transfers the ubiquitin to targeted substrates. Essential in early embryonic development to prevent apoptotic death.
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: G2/M phase-specific E3 ubiquitin-protein ligase; G2/M phase-specific HECT-type E3 ubiquitin transferase; G2/M-phase specific E3 ubiquitin ligase; PHD finger protein 7B
Gene Aliases: 6030408C04Rik; 9030416F18; AW046379; D930034K21Rik; G2E3; KIAA1333; mKIAA1333; PHF7B; RGD1310263
UniProt ID: (Human) Q7L622, (Mouse) Q5RJY2
Entrez Gene ID: (Human) 55632, (Rat) 299002, (Mouse) 217558
Molecular Function:
DNA metabolism protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support