Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: PDDSGTFYDQ AVVSNDMEEH LEEPVAEPEP DPEPEPEQEP VSEIQEEKPE PVLEETAPED AQKSSSPAPA DIAQTVQEDL RTFSWASVTS KNLPPSGAVP VTGIPPHVVK VPASQPRPES KPESQIPPQR PQRDQRVREQ RINIPPQRGP RPIREAGEQG DIEPRRMVRH PDSHQLFIGN LPHEVDKSEL KDFFQSYGNV VELRINSGGK LPNFGFVVFD DSEPVQKVLS NRPIMFRGEV RLNVEEKKTR AAREGDRRDN RLRGPGGPRG GLGGGMRGPP RGGMVQKPGF GVGRGLAPRQ
This gene encodes one of the DNA-unwinding enzymes which prefers partially unwound 3'-tailed substrates and can also unwind partial RNA/DNA and RNA/RNA duplexes in an ATP-dependent fashion. This enzyme is a member of the heterogeneous nuclear RNA-binding proteins and is also an element of the Ras signal transduction pathway. It binds specifically to the Ras-GTPase-activating protein by associating with its SH3 domain. Several alternatively spliced transcript variants of this gene have been described, but the full-length nature of some of these variants has not been determined.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ATP-dependent DNA helicase VIII; DNA helicase VIII; G3BP-1; GAP binding protein; GAP SH3 binding protein; GAP SH3 domain-binding protein 1; GTPase activating protein (SH3 domain) binding protein 1; hDH VIII; HDH-VIII; MGC111040; Ras GTPase-activating protein-binding protein 1; Ras-GTPase-activating protein SH3-domain binding protein 1; Ras-GTPase-activating protein SH3-domain-binding protein; RasGAP-associated endoribonuclease G3BP
Gene Aliases: AI849976; B430204O07; C87777; G3BP; G3BP1; HDH-VIII; mKIAA4115; RGD1310666
UniProt ID: (Human) Q13283, (Mouse) P97855
Entrez Gene ID: (Pig) 100513766, (Human) 10146, (Rat) 171092, (Mouse) 27041
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support