Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
G3BP2 gene ontology annotations related to this gene include stress granule assembly.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: G3BP-2; GAP SH3 domain-binding protein 2; GTPase activating protein (SH3 domain) binding protein 2; Ras GTPase-activating protein-binding protein 2; Ras-GTPase activating protein SH3 domain-binding protein 2
Gene Aliases: G3BP2; KIAA0660
UniProt ID: (Human) Q9UN86
Entrez Gene ID: (Human) 9908
Molecular Function:
RNA metabolism protein
nucleic acid metabolism protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support