Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: VVPVLLELSG PSGFLAPGSK VPLSLRIASF SGPQDLDLRT FVNPSFSLTS NLSRAHLELN ESAWGRLWLE VPDSAAPDSV VMVTVTAGGR EANPVPPTHA FLRLL
Highest antigen sequence identity to the following orthologs: Mouse - 86%, Rat - 79%.
VWA7 is a protein coding gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Protein G7c; von Willebrand factor A domain-containing protein 7
Gene Aliases: C6orf27; G7C; NG37; VWA7
UniProt ID: (Human) Q9Y334
Entrez Gene ID: (Human) 80737
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support