Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: STFAGHGRY AGHWTGDVWS SWEQLASSVP EILQFNLLGV PLVGADVCGF LGNTSEELCV RWTQLGAFYP FMRNHNSLLS LPQEPYSFSE PAQQAMRKAL TLRYALLPHL YTLFHQAHVA GETVARPLFL EFPKDSSTWT VDHQLLWGEA LLITPVLQAG KAEVTGYFPL GTWYDLQTVP VEALGSLPPP PAAPREPAIH SEGQWVTLPA PLDTINVHLR AGYIIPLQGP GLTTTESRQQ PMALAVALTK GGEARGELFW DDGESLEVLE RGAYTQVIFL ARNNTIVNEL VRVTSEGAGL QLQKVTVLGV ATAPQQVLSN GVPVSNFTYS PDTKVLDICV SLLMGEQFLV SWC (601-952 aa encoded by BC040431)
This gene encodes acid alpha-glucosidase, which is essential for the degradation of glycogen to glucose in lysosomes. Different forms of acid alpha-glucosidase are obtained by proteolytic processing. Defects in this gene are the cause of glycogen storage disease II, also known as Pompe's disease, which is an autosomal recessive disorder with a broad clinical spectrum. Three transcript variants encoding the same protein have been found for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: acid (Pompe disease, glycogen storage disease type II); acid alpha-glucosidase; Acid maltase; Aglucosidase alfa; glucosidase, alpha; acid (Pompe disease, glycogen storage disease type II); Lysosomal alpha-glucosidase
Gene Aliases: E430018M07Rik; GAA; LYAG
UniProt ID: (Human) P10253, (Mouse) P70699, (Rat) Q6P7A9
Entrez Gene ID: (Human) 2548, (Mouse) 14387, (Rat) 367562
Molecular Function:
glucosidase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support