Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Alomone Labs, LTD
FIGURE: 1 / 2
Reconstitution: 1 X 25 µL double distilled water (DDW), depending on the sample size. The antibody ships as a lyophilized powder at room temperature. Upon arrival, it should be stored at -20C. The reconstituted solution can be stored at 4C for up to 1 week. For longer periods, small aliquots should be stored at -20C. Avoid multiple freezing and thawing. Centrifuge all antibody preparations before use (10000 x g 5 min).
GAT-1 (SLC6A1) The protein encoded by SLC6A1 is a gamma-aminobutyric acid (GABA) transporter that localizes to the plasma membrane. The encoded protein removes GABA from the synaptic cleft, restoring it to presynaptic terminals.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: GABA T-1; GABA T1; GABA transporter 1; GABA transporter protein; GABAT1; gamma-aminobutyric acid (GABA-A) transporter 1; GAT-1; Sodium- and chloride-dependent GABA transporter 1; solute carrier family 6 (neurotransmitter transporter), member 1; solute carrier family 6 (neurotransmitter transporter, GABA), member 1; Solute carrier family 6 member 1
Gene Aliases: A730043E01; GABATHG; GABATR; Gabt; GABT1; GAT-1; GAT1; MAE; SLC6A1; XT-1; Xtrp1
UniProt ID: (Human) P30531, (Rat) P23978, (Mouse) P31648
Entrez Gene ID: (Human) 6529, (Rat) 79212, (Mouse) 232333
Molecular Function:
primary active transporter
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support