Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Sequence of this protein is as follows: MIITQTSHCY MTSLGILFLI NILPGTTGQG ESRRQEPGDF VKQDIGGLSP KHAPDIPDDS TDNITIFTRI LDRLLDGYDN RLRPGLGDAV TEVKTDIYVT SFGPVSDTDM EYTIDVFFRQ TWHDERLKFD GPMKILPLNN LLASKIWTPD TFFHNGKKSV AHNMTTPNKL LRLVDNGTLP YTMRLTIHAE CPMHLEDFPM DVHACPLKFG SYAYTTAEVV YSWTLGKNKS VEVAQDGSRL NQYDLLGHVV GTEIIRSSTG EYVVMTTHFH LKRKIGYFVI QTYLPCIMTV ILSQVSFWLN RESVPARTVF GVTTVLTMTT LSISARNSLP KVAYATAMDW FIAVCYAFVF SALIEFATVN YFTKRSWAWE GKKVPEALEM KKKTPAAPAK KTSTTFNIVG TTYPINLAKD TEFSTISKGA APSASSTPTI IASPKATYVQ DSPTETKTYN SVSKVDKISR IIFPVLFAIF NLVYWATYVN RESAIKGMIR KQ
GABA is the major inhibitory neurotransmitter in the mammalian brain where it acts at GABA-A receptors, which are ligand-gated chloride channels. Chloride conductance of these channels can be modulated by agents such as benzodiazepines that bind to the GABA-A receptor. At least 16 distinct subunits of GABA-A receptors have been identified.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: GABA(A) receptor subunit alpha-3; GABA(A) receptor, alpha 3; gamma-aminobutyric acid; gamma-aminobutyric acid (GABA) A receptor, alpha 3; Gamma-aminobutyric acid receptor subunit alpha-3; gamma-animobutyric acid (GABA) A receptor, alpha 3; MGC33793
Gene Aliases: GABRA3
UniProt ID: (Human) P34903
Entrez Gene ID: (Human) 2556
Molecular Function: ligand-gated ion channel
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support