Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
A 1:100 dilution of MA5-45746 was sufficient for detection of GABA-A R, Alpha5 in 20 µg of mouse brain lysate by ECL immunoblot analysis using Goat anti-mouse IgG:HRP as the secondary antibody. |Detects approximately 55kDa. Does not cross-react with GABA A Receptor Alpha2 or other GABA-A Receptors based on KO validation data.
This antibody was formerly sold as clone S415-24.
GABRA5 is a part of GABA, a major inhibitory neurotransmitter in the mammalian brain where it acts at GABA-A receptors, which are ligand-gated chloride channels. Chloride conductance of these channels can be modulated by agents such as benzodiazepines that bind to the GABA-A receptor. At least 16 distinct subunits of GABA-A receptors have been identified. Transcript variants utilizing three different alternative non-coding first exons have been described.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: GABA(A) receptor subunit alpha-5; GABA-A receptor alpha-5 subunit; GABAA receptor alpha5 subunit; gamma-aminobutyric acid; gamma-aminobutyric acid A receptor, alpha 5; gamma-aminobutyric acid (GABA) A receptor, alpha 5; gamma-aminobutyric acid (GABA-A) receptor, subunit alpha 5; gamma-aminobutyric acid A receptor, alpha 5; Gamma-aminobutyric acid receptor subunit alpha-5; ligand-gated ion channel subunit
Gene Aliases: A230018I05Rik; Gabra-5; GABRA5
UniProt ID: (Human) Q9NYT4, (Rat) P19969, (Mouse) Q8BHJ7
Entrez Gene ID: (Human) 2558, (Rat) 29707, (Mouse) 110886
Molecular Function:
ligand-gated ion channel
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support