Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Immunogen sequence: PYVKAIDMYL MGCFVFVFMA LLEYALVNYI FFGRGPQRQK KAAEKAASAN NEKMRLDVNK IFYKDIKQNG TQYRSLWDPT GNLSPTRRTT NYDFSLYTMD P
GAD-65 and GAD-67, glutamate decarboxylases, function to catalyze the production of GABA (gamma-aminobutyric acid). In the central nervous system GABA functions as the main inhibitory transmitter by increasing a Cl- conductance that inhibits neuronal firing. GABA has been shown to activate both ionotropic (GABAA) and metabotropic (GABAB) receptors as well as a third class of receptors called GABAC. Both GABAA and GABAC are ligand-gated ion channels, however, they are structurally and functionally distinct. Members of the GABAA receptor family include GABAA Rα1-6, GABAA R β1-3, GABAA Rγ1-3, GABAA Rδ, GABAA Rε, GABAA Rρ1 and GABAA Rρ2. The GABAB family is composed of GABAB R1α and GABAB R1β. GABA transporters have also been identified and include GABA T-1, GABA T-2 and GABA T-3 (also designated GAT-1, -2, and -3). The GABA transporters function to terminate GABA action.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: GABA(A) receptor subunit beta-2; GABAARbeta2; gamma-aminobutyric acid; gamma-aminobutyric acid (GABA-A) receptor, subunit beta 2; gamma-aminobutyric acid A receptor beta 2; gamma-aminobutyric acid A receptor, subunit beta 2; Gamma-aminobutyric acid receptor beta 2; gamma-aminobutyric acid receptor beta-2 subunit; Gamma-aminobutyric acid receptor subunit beta-2; gamma-aminobutyric acid receptor, subunit beta 2; MGC119386; MGC119388; MGC119389
Gene Aliases: AI834970; C030002O17Rik; C030021G16Rik; Gabrab2; Gabrb-2; GABRB2; GARB2
UniProt ID: (Human) P47870, (Mouse) P63137, (Rat) P63138
Entrez Gene ID: (Human) 2561, (Mouse) 14401, (Rat) 25451
Molecular Function:
ligand-gated ion channel
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support