Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Cell lysate).
Immunogen sequence: MLSKVLPVFL GILLILQSRV EGPQTESKNE ASSRDVVYGP QPQPLENQLL SEETKSTETE TGSRVGKLPE ASRILNTILS NYDHKLRPGI GEKPTVVTVE
The product of this gene belongs to the ligand-gated ionic channel (TC 1.A.9) family. It encodes the gamma-aminobutyric acid (GABA) A receptor which is a multisubunit chloride channel that mediates the fastest inhibitory synaptic transmission in the central nervous system. This gene encodes an epsilon subunit. It is mapped to chromosome Xq28 in a cluster comprised of genes encoding alpha 3, beta 4 and theta subunits of the same receptor. Alternatively spliced transcript variants have been identified, but only one is thought to encode a protein.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: GABA(A) mu Receptor; GABA(A) receptor subunit epsilon; GABA(A) receptor, epsilon; gamma-aminobutyric acid; gamma-aminobutyric acid (GABA) A receptor, epsilon; Gamma-aminobutyric acid receptor subunit epsilon
Gene Aliases: GABRE
UniProt ID: (Human) P78334
Entrez Gene ID: (Human) 2564
Molecular Function:
ligand-gated ion channel
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support