Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
The gamma-aminobutyric acid (GABA) A receptor is a multisubunit chloride channel that mediates the fastest inhibitory synaptic transmission in the central nervous system. This gene encodes GABA A receptor, theta subunit. GABRQ gene is mapped to chromosome Xq28 in a cluster including the genes encoding the alpha 3 and epsilon subunits of the same receptor. This gene location is also the candidate region of 2 different neurologic diseases:early-onset parkinsonism (Waisman syndrome) and X-linked mental retardation (MRX3).The gamma-aminobutyric acid (GABA) A receptor is a multisubunit chloride channel that mediates the fastest inhibitory synaptic transmission in the central nervous system. This gene encodes GABA A receptor, theta subunit. It is mapped to chromosome Xq28 in a cluster including the genes encoding the alpha 3 and epsilon subunits of the same receptor. This gene location is also the candidate region of 2 different neurologic diseases: early-onset parkinsonism (Waisman syndrome) and X-linked mental retardation (MRX3).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: GABA(A) receptor subunit theta; GABA(A) Theta; gamma-aminobutyric acid; gamma-aminobutyric acid A receptor, theta; gamma-aminobutyric acid (GABA) A receptor, theta; gamma-aminobutyric acid (GABA) receptor, theta; gamma-aminobutyric acid (GABA-A) receptor, subunit theta; gamma-aminobutyric acid A receptor, theta; Gamma-aminobutyric acid receptor subunit theta
Gene Aliases: GABRQ; THETA
UniProt ID: (Human) Q9UN88, (Mouse) Q9JLF1
Entrez Gene ID: (Human) 55879, (Rat) 65187, (Mouse) 57249
Molecular Function:
ligand-gated ion channel
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support