Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAD-65 and GAD-67 are members of the group II decarboxylase family of proteins and are responsible for catalyzing the rate limiting step in the production of GABA (gamma-aminobutyric acid) from L-glutamic acid. Although both GADs are found in the brain, GAD-65 localizes to synaptic vesicle membranes in nerve terminals, while GAD-67 is distributed throughout the cell. GAD-67 is responsible for the basal levels of GABA synthesis. In the case of a heightened demand for GABA in neurotransmission, GAD-65 will transiently activate to assist in GABA production. The loss of GAD-65 is detrimental and can impair GABA neurotransmission, however the loss of GAD-67 is lethal. Due to alternative splicing, two isoforms exist for GAD-67: the predominant GAD-67 form and the minor GAD-25 form.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 67 kDa glutamic acid decarboxylase; FLJ45882; GAD 67; GAD-67; Glutamate decarboxylase 1; glutamate decarboxylase 1 (brain, 67kDa); Glutamate decarboxylase 67 kDa isoform; OTTHUMP00000041055
Gene Aliases: CPSQ1; GAD; GAD1; GAD67; SCP
UniProt ID: (Human) Q99259
Entrez Gene ID: (Human) 2571
Molecular Function:
decarboxylase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support