Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
This Antibody was verified by Relative expression to ensure that the antibody binds to the antigen stated.
FIGURE: 1 / 3
A suggested positive control for immunohistochemical applications is brain. In Western blot, a doublet at ~65/67 kDa has been observed.
Rabbit anti Glutamate Decarboxylase 1/2 antibody recognizes both molecular forms of glutamate decarboxylase, GAD1 (GAD67) and GAD2 (GAD65), the enzyme which converts glutamic acid to gamma -aminobutyric acid (GABA), the major inhibitory transmitter in the higher brain region.
GAD-65 and GAD-67 are members of the group II decarboxylase family of proteins and are responsible for catalyzing the rate limiting step in the production of GABA ( gamma-aminobutyric acid) from L-glutamic acid. Although both GADs are found in the brain, GAD-65 localizes to synaptic vesicle membranes in nerve terminals, while GAD-67 is distributed throughout the cell. GAD-67 is responsible for the basal levels of GABA synthesis. In the case of a heightened demand for GABA in neurotransmission, GAD-65 will transiently activate to assist in GABA production. The loss of GAD-65 is detrimental and can impair GABA neurotransmission, however the loss of GAD-67 is lethal. Due to alternative splicing, two isoforms exist for GAD-67: the predominant GAD-67 form and the minor GAD-25 form.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 65 kDa glutamic acid decarboxylase; 67 kDa glutamic acid decarboxylase; EC 4.1.1.15; FLJ45882; GAD 65; GAD 67; GAD-65; GAD-67; Glutamate decarboxylase 1; Glutamate decarboxylase 1 (brain); glutamate decarboxylase 1 (brain, 67kDa); glutamate decarboxylase 1 variant GAD67NT; Glutamate decarboxylase 2; Glutamate decarboxylase 2 (islet); glutamate decarboxylase 2 (pancreatic islets and brain, 65kDa); Glutamate decarboxylase 65 kDa isoform; Glutamate decarboxylase 67 kDa isoform; GLUTAMATE DECARBOXYLASE, 67 KD ISOFORM (GAD-67) (67 KD GLUTAMIC ACID DECARBOXYLASE); Glutamate decarboxylase-2 (pancreas); glutamic acid decarboxylase; glutamic acid decarboxylase 1; glutamic acid decarboxylase 2; glutamic acid decarboxylase 65
Gene Aliases: 6330404F12Rik; CPSQ1; EP10; GAD; GAD(65); Gad-1; Gad-2; GAD1; GAD2; GAD25; GAD44; GAD65; GAD67; SCP
UniProt ID: (Human) Q99259, (Cat) P14748, (Human) Q05329, (Mouse) O08685, (Rat) P18088, (Mouse) P48320, (Rat) Q05683
Entrez Gene ID: (Human) 2571, (Cat) 493699, (Human) 2572, (Mouse) 14415, (Rat) 24379, (Mouse) 14417, (Rat) 24380
Molecular Function:
decarboxylase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support