Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: SDEEEGEVKA LGAAEKDGEA ECPPCIPPPS AFLKAWVYWP GEDTEEEEDE EEDEDSDSGS DEEEGEAEAS SSTPATGVFL KSWVYQPGED TEEEEDEDSD TGSAEDEREA ETSASTPPAS AFLKAWVYRP GEDTEEEEDE DVDSEDKEDD SEAALGEAES DPHPSHPDQR AHFRGWGYRP GKETEEEEAA EDWGEAEPCP FRVAIYVPGE KPPPPWAPPR LPLRLQRRLK RPETPTHDPD PETPLKARKV RFSEKVTVHF LAVWAG
This gene is a member of a group of genes whose transcript levels are increased following stressful growth arrest conditions and treatment with DNA-damaging agents. The induction of this gene by ionizing radiation occurs in certain cell lines regardless of p53 status, and its protein response is correlated with apoptosis following ionizing radiation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Growth arrest and DNA damage-inducible protein GADD34; growth arrest and DNA-damage-inducible 34; myeloid differentiation primary response gene 116; Myeloid differentiation primary response protein MyD116; Myeloid differentiation primary response protein MyD116 homolog; PR15A; Protein phosphatase 1 regulatory subunit 15A; protein phosphatase 1, regulatory (inhibitor) subunit 15A; protein phosphatase 1, regulatory subunit 15A
Gene Aliases: 9630030H21; GADD34; Myd116; PPP1R15A
UniProt ID: (Human) O75807, (Mouse) P17564
Entrez Gene ID: (Human) 23645, (Mouse) 17872
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support