Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
GAGE12F is a protein coding gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: AL4; Cancer/testis antigen 4.7; CT4.7; G antigen 12F; g antigen 12F/G/I; G antigen 12G; G antigen 12I; G antigen 7; GAGE-12F; GAGE-12G; GAGE-12I; GAGE-7; GAGE-7B; GAGE-8
Gene Aliases: GAGE12F; GAGE12G; GAGE12I; GAGE7; GAGE7B
UniProt ID: (Human) P0CL81
Entrez Gene ID: (Human) 100008586
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support