Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Recognizes GAL4 derivatives containing the C-terminal activation domain.
GAL4 is a transcriptional activator which activates genes necessary for galactose catabolism. The DNA binding domain of GAL4 is contained in the first 74 amino acids and contains a "zinc finger" sequence motif. However, amino acids 1-147 have been shown to bind to DNA in vitro. While GAL4 (1-74) bind DNA specifically, it is with a lower affinity than the GAL4 (1-147). GAL4 (1-147) binds to a specific 17 base pair nucleotide sequence, but alone is insufficient to activate transcription. Either of the two acidic activating regions, activating region I (amino acids 148-238) or activating region II (amino acids 768-881), must be joined to activate transcription.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Regulatory protein GAL4
Gene Aliases: GAL4; GAL81; YPL248C
UniProt ID: (Baker's yeast) P04386
Entrez Gene ID: (Baker's yeast) 855828
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support