Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Peptide Sequence: RLQTAVAELR ARREALAKHC LVGGEASDPK YITDRRFRPF QFGSAKVLGY ILRSGLSPQD QEECERLATP ELQYKDKLDA KQFPPTVSLP LKTSRPLSP
Catalyzes the transfer of sulfate to beta-1,3-linked galactose residues in O-linked glycoproteins. Good substrates include asialofetuin, Gal-beta-1,3-GalNAc and Gal-beta-1,3 (GlcNAc-beta-1,6)GalNAc.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: beta-galactose-3-O-sulfotransferase; Beta-galactose-3-O-sulfotransferase 4; beta-galactose-3-O-sulfotransferase, 4; Gal-beta-1,3-GalNAc 3'-sulfotransferase; Gal3ST-4; Galactose-3-O-sulfotransferase 4; galbeta1-3GalNAc 3'-sulfotransferase
Gene Aliases: GAL3ST-4; GAL3ST4; PP6968
UniProt ID: (Human) Q96RP7
Entrez Gene ID: (Human) 79690
Molecular Function: transferase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support