Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MAALRQPQV AELLAEARRA FREEFGAEPE LAVSAPGRVN LIGEHTDYNQ GLVLPMALEL MTVLVGSPRK DGLVSLLTTS EGADEPQRLQ FPLPTAQRSL EPGTPRWANY VKGVIQYYPA APLPGFSAVV VSSVPLGGGL SSSASLEVAT YTFLQQLCPD SGTIAARAQV CQQAEHSFAG MPCGIMDQFI SLMGQKGHAL LIDCRSLETS LVPLSDPKLA VLITNSNVRH SLASSEYPVR RRQCEEVARA LGKESLREVQ LEELEAARDL VSKEGFRRAR HVVGEIRRTA QAAAALRRGD YRAFGRLMVE SHRSLRDDYE VSCPELDQLV EAALAVPGVY GSRMTGGGFG GCTVTLLEAS AAPHAMRHIQ EHYGGTATFY LSQAADGAKV LCL (1-392 aa encoded by BC001166)
Galactokinase is a major enzyme for the metabolism of galactose and its deficiency causes congenital cataracts in the adult population. GALK1 sequence shares the greatest level of conservation, 44. 5% identity with that from E. coli and 34. 6% amino acid identity with the product of the human GALK2 gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: epididymis secretory protein Li 19; Galactokinase; Galactose kinase
Gene Aliases: AA409894; GALK; GALK1; Gk; GK1; Glk; Glk1; HEL-S-19
UniProt ID: (Human) P51570, (Mouse) Q9R0N0
Entrez Gene ID: (Human) 2584, (Mouse) 14635
Molecular Function:
carbohydrate kinase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support