Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Sequence of this protein is as follows: PCGIMDQFIS LMGQKGHALL IDCRSLETSL VPLSDPKLAV LITNSNVRHS LASSEYPVRR RQCEEVARAL GKESLREVQL EELEAARDLV SKEGFRRAR
Galactokinase is a major enzyme for the metabolism of galactose and its deficiency causes congenital cataracts in the adult population. GALK1 sequence shares the greatest level of conservation, 44. 5% identity with that from E. coli and 34. 6% amino acid identity with the product of the human GALK2 gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: epididymis secretory protein Li 19; Galactokinase; Galactose kinase
Gene Aliases: GALK; GALK1; GK1; HEL-S-19
UniProt ID: (Human) P51570
Entrez Gene ID: (Human) 2584
Molecular Function: carbohydrate kinase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support