Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
This gene encodes a highly efficient N-acetylgalactosamine kinase, which has galactokinase activity when galactose is present at high concentrations. Two alternatively spliced transcript variants encoding different isoforms have been found for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Galactokinase 2; GalNAc kinase; N-acetylgalactosamine kinase
Gene Aliases: 2810017M24Rik; AI327030; GALK2; GK2
UniProt ID: (Human) Q01415, (Mouse) Q68FH4, (Rat) Q5XIG6
Entrez Gene ID: (Human) 2585, (Mouse) 69976, (Rat) 296117
Molecular Function:
carbohydrate kinase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support