Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MRRLTRRLVL PVFGVLWITV LLFFWVTKRK LEVPTGPEVQ TPKPSDADWD DLWDQFDERR YLNAKKWRVG DDPYKLYAFN QRESERISSN RAIPDTRHLR CTLLVYCTDL PPTSIIITFH NEARSTLLRT IRSVLNRTPT HLIREIILVD DFSNDPDDCK QLIKLPKVKC LRNNERQGLV RSRIRGADIA QGTTLTFLDS HCEVNRDWLQ PLLHRVKEDY TRVVCPVIDI INLDTFTYIE SASELRGGFD WSLHFQWEQL SPEQKARRLD PTEPIRTPII AGGLFVIDKA WFDYLGKYDM DMDIWGGENF EISFRVWMCG GSLEIVPCSR VGHVFRKKHP YVFPDGNANT YIKNTKRTAE VWMDEYKQYY YAARPFALER PFGNVESRLD LRKNLRCQSF KWYLENIYPE LSIPKESSIQ KGNIRQRQKC LESQRQNNQE TPNLKLSPCA KVKGEDAKSQ VWAFTYTQQI LQEELCLSVI TLFPGAPVVL VLCKNGDDRQ QWTKTGSHIE HIASHLCLDT DMFGDGTENG KEIVVNPCES SLMSQHWDMV SS
GALNT14 (EC 2.4.1.41) belongs to a large subfamily of glycosyltransferases residing in the Golgi apparatus. GALNT enzymes catalyze the first step in the O-glycosylation of mammalian proteins by transferring N-acetyl-D-galactosamine (GalNAc) to peptide substrates.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: FLJ12691; FLJ13977; GalNAc-T14; Polypeptide GalNAc transferase 14; Polypeptide N-acetylgalactosaminyltransferase 14; polypeptide N-acetylgalactosaminyltransferase 15; pp-GaNTase 14; Protein-UDP acetylgalactosaminyltransferase 14; UDP-GalNAc:polypeptide N-acetylgalactosaminyltransferase 14; UDP-GalNAc:polypeptide N-acetylgalactosaminyltransferase T10; UDP-N-acetyl-alpha-D-galactosamine:polypeptide N-acetylgalactosaminyltransferase 14 (GalNAc-T14)
Gene Aliases: GalNac-T10; GalNac-T14; GALNT14; GALNT15; UNQ2434/PRO4994
UniProt ID: (Human) Q96FL9
Entrez Gene ID: (Human) 79623
Molecular Function:
glycosyltransferase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support