Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MAHLKRLVK LHIKRHYHKK FWKLGAVIFF FIIVLVLMQR EVSVQYSKEE SRMERNMKNK NKMLDLMLEA VNNIKDAMPK MQIGAPVRQN IDAGERPCLQ GYYTAAELKP VLDRPPQDSN APGASGKAFK TTNLSVEEQK EKERGEAKHC FNAFASDRIS LHRDLGPDTR PPEYVEE (1-176 aa encoded by BC056246)
GALNT3 encodes UDP-GalNAc transferase 3, a member of the GalNAc-transferases family. This family transfers an N-acetyl galactosamine to the hydroxyl group of a serine or threonine residue in the first step of O-linked oligosaccharide biosynthesis. Individual GalNAc-transferases have distinct activities and initiation of O-glycosylation is regulated by a repertoire of GalNAc-transferases. The protein is highly homologous to other family members, however the enzymes have different substrate specificities.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: GalNAc transferase 3; GalNAc-T3; Polypeptide GalNAc transferase 3; polypeptide GalNAc-transferase T3; Polypeptide N-acetylgalactosaminyltransferase 3; pp-GaNTase 3; ppGaNTase-T3; Protein-UDP acetylgalactosaminyltransferase 3; UDP-GalNAc:polypeptide N-acetylgalactosaminyltransferase 3; UDP-N-acetyl-alpha-D-galactosamine:polypeptide N-acetylgalactosaminyltransferase 3 (GalNAc-T3)
Gene Aliases: GalNAc-T3; GALNT3; HFTC; HHS
UniProt ID: (Human) Q14435, (Mouse) P70419
Entrez Gene ID: (Human) 2591, (Rat) 366061, (Mouse) 14425
Molecular Function:
glycosyltransferase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support