Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MAVARKIRT LLTVNILVFV GIVLFSVYCR LQGRSQELVR IVSGDRRVRS RHAKVGTLGD REAILQRLDH LEEVVYNQLN GLAKPIGLVE GPGGLGQGGL AATLRDDGQE AEGKYEEYGY NAQLSDRISL DRSIPDYRPR KCRQMSYAQD LPQVSV (1-155 aa encoded by B C157845)
This gene encodes a member of the UDP-N-acetyl-alpha-D-galactosamine:polypeptide N-acetylgalactosaminyltransferase (GalNAc-T) family of enzymes. GalNAc-Ts initiate mucin-type O-linked glycosylation in the Golgi apparatus by catalyzing the transfer of GalNAc to serine and threonine residues on target proteins. They are characterized by an N-terminal transmembrane domain, a stem region, a lumenal catalytic domain containing a GT1 motif and Gal/GalNAc transferase motif, and a C-terminal ricin/lectin-like domain. GalNAc-Ts have different, but overlapping, substrate specificities and patterns of expression. This gene is expressed specifically in the brain, with highest expression in the cerebellum. Multiple transcript variants encoding different isoforms have been found for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: GalNAc transferase 9; GalNAc-T9; Polypeptide GalNAc transferase 9; Polypeptide N-acetylgalactosaminyltransferase 9; pp-GaNTase 9; Protein-UDP acetylgalactosaminyltransferase 9; UDP-GalNAc: polypeptide N-acetylgalactosaminyltransferase 9; UDP-GalNAc:polypeptide N-acetylgalactosaminyltransferase 9; UDP-N-acetyl-alpha-D-galactosamine:polypeptide N-acetylgalactosaminyltransferase 9 (GalNAc-T9)
Gene Aliases: GALNAC-T9; GALNACT9; GALNT9
UniProt ID: (Human) Q9HCQ5
Entrez Gene ID: (Human) 50614, (Rat) 304571, (Mouse) 231605
Molecular Function:
glycosyltransferase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support