Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: DKHLVKSAEP GEQQTFPLGL GDGQFYSWTD GLRRKDWHDY ESIQKEAMRS GKGEHGKPYP LTEEDHDDSA YRENGFNIFV SN
Highest antigen sequence identity to the following orthologs: Mouse - 93%, Rat - 37%.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: GalNAc transferase 17; GalNAc-T17; GaNTase 17; Polypeptide GalNAc transferase 17; polypeptide GalNAc transferase-like 6; Polypeptide N-acetylgalactosaminyltransferase-like 6; pp-GaNTase 17; Protein-UDP acetylgalactosaminyltransferase 17; Putative polypeptide N-acetylgalactosaminyltransferase 17; UDP-GalNAc:polypeptide N-acetylgalactosaminyltransferase 17; UDP-N-acetyl-alpha-D-galactosamine:polypeptide N-acetylgalactosaminyltransferase 20; UDP-N-acetyl-alpha-D-galactosamine:polypeptide N-acetylgalactosaminyltransferase-like 6
Gene Aliases: GalNAc-T6L; GALNACT20; GALNT17; GALNTL6
UniProt ID: (Human) Q49A17
Entrez Gene ID: (Human) 442117
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support