Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
This antibody is predicted to react with bovine, canine, human and mouse based on sequence homology.
This antibody is tested in Peptide ELISA: antibody detection limit dilution 128,000.
The neuropeptide galanin elicits a range of biological effects by interaction with specific G-protein-coupled receptors. Galanin receptors are seven-transmembrane proteins shown to activate a variety of intracellular second-messenger pathways. GALR1 inhibits adenylyl cyclase via a G protein of the Gi/Go family. GALR1 is widely expressed in the brain and spinal cord, as well as in peripheral sites such as the small intestine and heart.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Gal receptor 1; Gal-1 receptor; Gal-r1; GAL1-R; Galanin r1 receptor; Galanin receptor type 1; Galanin-1 receptor; Galn1r; GALR-1; Hugal 1-r
Gene Aliases: Galnr; Galnr1; Galr1
UniProt ID: (Rat) Q62805
Entrez Gene ID: (Rat) 50577
Molecular Function:
G-protein coupled receptor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support