Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: VNPIVYALVS KHFRKGFRTI CAGLLGRAPG RASGRVCAAA RGTHSGSVLE RESSDLLHMS EAAGALRPCP GASQPCILEP CPGPSWQGPK AGDSILTVDV A
GalR2 expression has been documented widely in the brain and in peripheral tissues including small intestine, heart, kidney, and liver. Localization data from rat CNS indicate high levels of expression in dorsal root ganglia, amygdala, and Purkinje neurons. G-protein Coupled Receptors (GPCRs) comprise one of the largest families of signaling molecules with more than a thousand members currently predicted to exist. All GPCRs share a structural motif consisting of seven membrane-spanning helices, and exist in both active and inactive forms. An array of activating ligands participate in the conformation of GPCRs which leads to signaling via G-proteins and downstream effectors. Ongoing studies have also shown the vast series of reactions which participate in the negative regulation of GPCRs. This "turn-off" activity has tremendous implications for the physiological action of the cell, and continues to drive pharmacological research for new drug candidates. Two blockbuster drugs which have been developed as GPCR-targeted pharmaceuticals are Zyprexa (Eli Lilly) and Claritin (Schering-Plough) which have multi-billion dollar shares of the mental health and allergy markets, respectively.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Gal receptor 2; Gal-2 receptor; Gal-r2; GAL2-R; Galanin r2 receptor; Galanin receptor type 2; Galanin-2 receptor; MGC125983; MGC125984
Gene Aliases: GAL2-R; GALNR2; GALR-2; GALR2
UniProt ID: (Human) O43603
Entrez Gene ID: (Human) 8811
Molecular Function:
G-protein coupled receptor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support