Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MSAPSATPI FAPGENCSPA WGAAPAAYDA ADTHLRILGK PVMERWETPY MHALAAAASS KGGRVLEVGF GMAIAASKVQ EAPIDEHWII ECNDGVFQRL RDWAPRQTHK VIPLKGLWED VAPTLPDGHF DGILYDTYPL SEETWHTHQF NFIKNHAFRL LKPGGVLTYC NLTSWGELMK SKYSDITIMF EETQVPALLE AGFRRENIRT EVMALVPPAD CRYYAFPQMI TPLVTKG (1-236 aa encoded by BC016760)
GAMT is a methyltransferase that converts guanidoacetate to creatine, using S-adenosylmethionine as the methyl donor. Defects in its gene have been implicated in neurologic syndromes and muscular hypotonia, probably due to creatine deficiency and accumulation of guanidinoacetate in the brain of affected individuals.The protein encoded by this gene is a methyltransferase that converts guanidoacetate to creatine, using S-adenosylmethionine as the methyl donor. Defects in this gene have been implicated in neurologic syndromes and muscular hypotonia, probably due to creatine deficiency and accumulation of guanidinoacetate in the brain of affected individuals. Two transcript variants encoding different isoforms have been described for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: epididymis secretory protein Li 20; guanidinoacetate methyltransferase; Guanidinoacetate N-methyltransferase
Gene Aliases: AA571402; CCDS2; GAMT; GMT; HEL-S-20; PIG2; Spintz1; TP53I2
UniProt ID: (Human) Q14353, (Mouse) O35969
Entrez Gene ID: (Human) 2593, (Pig) 100625146, (Mouse) 14431, (Rat) 25257
Molecular Function:
methyltransferase
transferase
metabolite interconversion enzyme
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support