Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: DLDTGTNYDY VREFKRSTGT WHHTKPLLPS DLRRTGCAAL RIANCKLFRL QLQQGLFRIR VHSP
This gene encodes a member of the cytoskeletal BTB/kelch repeat family. The encoded protein plays a role in neurofilament architecture and is involved in mediating the ubiquitination and degradation of some proteins. Defects in this gene are a cause of giant axonal neuropathy.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Gigaxonin; kelch-like family member 16; Kelch-like protein 16
Gene Aliases: GAN; GAN1; KLHL16
UniProt ID: (Human) Q9H2C0
Entrez Gene ID: (Human) 8139
Molecular Function:
scaffold/adaptor protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support