Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Glycosyl hydrolase enzymes hydrolyse the glycosidic bond between two or more carbohydrates, or between a carbohydrate and a non-carbohydrate moiety. This gene encodes a member of glycosyl hydrolases family 31. This enzyme hydrolyses terminal, non-reducing 1,4-linked alpha-D-glucose residues and releases alpha-D-glucose. This is a key enzyme in glycogen metabolism and its gene localizes to a chromosomal region (15q15) that is associated with susceptibility to diabetes.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: glucosidase, alpha; neutral C; Neutral alpha-glucosidase C
Gene Aliases: 5830445O15Rik; 9330160A12; GANC; mFLJ00088
UniProt ID: (Human) Q8TET4, (Mouse) Q8BVW0
Entrez Gene ID: (Human) 2595, (Mouse) 76051, (Rat) 24382
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support