Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAPVD1 acts both as a GTPase-activating protein (GAP) and a guanine nucleotide exchange factor (GEF), and participates in various processes such as endocytosis, insulin receptor internalization or LC2A4/GLUT4 trafficking. It acts as a GEF for the Ras-related protein RAB31 by exchanging bound GDP for free GTP, leading to regulate LC2A4/GLUT4 trafficking. In the absence of insulin, it maintains RAB31 in an active state and promotes a futile cycle between LC2A4/GLUT4 storage vesicles and early endosomes, retaining LC2A4/GLUT4 inside the cells. Upon insulin stimulation, it is translocated to the plasma membrane, releasing LC2A4/GLUT4 from intracellular storage vesicles. It is also involved in EGFR trafficking and degradation, possibly by promoting EGFR ubiquitination and subsequent degradation by the proteasome. It has GEF activity for Rab5 and GAP activity for Ras.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: DKFZP434C212; GAPex-5; GTPase activating protein and VPS9 domains 1; GTPase-activating protein and VPS9 domain-containing protein 1; MGC138847; MGC138848; Rab5-activating protein 6
Gene Aliases: 2010005B09Rik; 4432404J10Rik; AW108497; GAPex-5; GAPEX5; GAPVD1; KIAA1521; mKIAA1521; RAP6; RGD1307479; RME-6
UniProt ID: (Human) Q14C86, (Mouse) Q6PAR5
Entrez Gene ID: (Human) 26130, (Mouse) 66691, (Rat) 311880
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support