Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MCTALSPKV RSGPGLSDMH QYSQWLASRH EANLLPMKED LALWLTNLLG KEITAETFME KLDNGALLCQ LAETMQEKFK ESMDANKPTK NLPLKKIPCK TSAPSGSFFA RDNTANFLSW CRDLGVDETC LFESEGLVLH KQPREVCLCL LELGRIAARY GVEPPGLIKL EKEIEQEETL SAPSPSPSPS SKSSGKKSTG NLLDDAVKRI SEDPPCKCPN KFCVERLSQG RYRVGEKILF IRMLHNKHVM VRVGGGWETF AGYLLKHDPC RMLQISRVDG KTSPIQSKSP TLKDMNPDNY LVVSASYKAK KEIK (1-313 aa encoded by BC040470)
Gas2 is a 313 amino acid protein encoded by the human gene GAS2. Gas2 is thought to play a role in apoptosis by acting as a cell death substrate for caspases. Gas2, a component of the microfilament system, is cleaved by a caspase (caspase-3 and caspase-7) at Asparagine 278 during apoptosis. The cleaved form resulting from this dramatically induces the rearrangement of the Actin cytoskeleton and causes potent changes in the shape of the affected cells. Gas2 is believed to also be involved in the membrane ruffling process. During the G0-G1 transition phase Gas2 can be found phosphorylated on its serine residues. Gas2 is a cytoskeleton and peripheral membrane protein that co-localizes with Actin fibers at the cell border and along the stress fibers in growth-arrested fibroblasts. Gas2 is mainly membrane-associated but when hyperphosphorylated it will accumulate at membrane ruffles. Gas2 is specifically expressed at growth arrest and is ubiquitously expressed with highest levels found in liver, lung and kidney.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: GAS 2; GAS-2; growth arrest-specific 2; Growth arrest-specific protein 2
Gene Aliases: Gas-2; GAS2; RGD1563167
UniProt ID: (Human) O43903, (Mouse) P11862
Entrez Gene ID: (Human) 2620, (Rat) 499156, (Mouse) 14453
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support