Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
This Antibody was verified by Knockdown to ensure that the antibody binds to the antigen stated.
FIGURE: 1 / 9
Recommended positive controls: HepG2 nuclear extract, mouse heart.
Predicted reactivity: Mouse (89%), Rat (89%), Xenopus laevis (82%), Dog (96%), Pig (93%), Chicken (85%), Bovine (95%).
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
GATA4 is a member of the GATA family of zinc-finger transcription factors. Members of this family recognize the GATA motif which is present in the promoters of many genes. This protein is thought to regulate genes involved in embryogenesis and in myocardial differentiation and function. Mutations in the gene encoding GATA4 have been associated with cardiac septal defects.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: DNA-binding protein GATA-GT2; GAT4; GATA-binding factor 4; GATA-binding protein 4; MGC126629; Transcription factor GATA-4
Gene Aliases: ASD2; Gata-4; GATA4; TACHD; TOF; VSD1
UniProt ID: (Human) P43694, (Mouse) Q08369, (Rat) P46152
Entrez Gene ID: (Human) 2626, (Mouse) 14463, (Rat) 54254
Molecular Function:
DNA-binding transcription factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support