Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Positive test controls include: HeLa. The target is usually found in the following locations: Cytoplasm, Endoplasmic reticulum-Golgi intermediate compartment, Golgi apparatus, Lipid droplet, Membrane, Peripheral membrane protein, cis-Golgi network, trans-Golgi network.
Immunogen sequence: MVDKNIYIIQ GEINIVVGAI KRNARWSTHT PLDEERDPLL HSFGHLKEVL NSITELSEIE PNVFLRPFLE VIRSEDTTGP ITGLA
This gene encodes a member of the Sec7 domain family. The encoded protein is a guanine nucleotide exchange factor that regulates the recruitment of proteins to membranes by mediating GDP to GTP exchange. The encoded protein is localized to the Golgi apparatus and plays a role in vesicular trafficking by activating ADP ribosylation factor 1. The encoded protein has also been identified as an important host factor for viral replication. Multiple transcript variants have been observed for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: BFA-resistant GEF 1; Golgi-specific brefeldin A-resistance guanine nucleotide exchange factor 1
Gene Aliases: 1700083E03Rik; AI035702; ARF1GEF; AW049900; GBF1; KIAA0248; mKIAA0248
UniProt ID: (Human) Q92538, (Mouse) Q6DFZ1
Entrez Gene ID: (Human) 8729, (Mouse) 107338
Molecular Function:
guanyl-nucleotide exchange factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support