Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: VHRRKLAQLE KLQDEELDPE FVQQVADFCS YIFSNSKTKT LSGGIQVNGP RLESLVLTYV NAISSGDLPC MENAVLALAQ IENSAAVQKA IAHYEQQMGQ KVQLPTESLQ ELLDLHRDSE REAIEVFIRS SFKDVDHLFQ KELAAQLEKK RDDFCKQNQE ASSDRCSGLL QVIFSPLEEE VKAGIYSKPG GYRLFVQKLQ DLKKKYYEEP RKGIQAEEIL QTYLKSKESM TDAILQTDQT LTEKEKEIEV ERVKAESAQA SAKMLQEMQR KNEQMMEQKE RSYQEHLKQL TEKMENDRVQ LLKEQERTLA LKLQEQEQLL KEGFQKESRI MKNEIQDLQT KMRRRKACTI S
GBP1 or guanlyate binding protein 1 is a 68 kDa GTPase and member of the GTPase1 family of molecules. GBP1 expression is induced by Interferon g as well as other inflammatory cytokines including IL-1a, IL-1b and TNF-a. Induction of GBP1 expression by poly I:C, a prototypical TLR3 ligand, as well as GBP1 expression dependency on NF-kB and IFNa response elements implicate TLR pathway involvement. Inversely proportional expression of GBP1 and MMP-1 as well as other related molecules to adhesion and migration show a clear functional role of GBP1 in suppressing angiogenesis and migration especially of endothelial cells. As a regulatory molecule it is centrally involved in signal transduction of inflammatory and cell migration networks.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: GBP-1; GTP-binding protein 1; Guanine nucleotide-binding protein 1; guanylate binding protein 1, interferon-inducible; guanylate binding protein 1, interferon-inducible, 67kDa; Guanylate-binding protein 1; huGBP-1; Interferon-induced guanylate-binding protein 1
Gene Aliases: GBP1
UniProt ID: (Human) P32455
Entrez Gene ID: (Pig) 100151938, (Human) 2633, (Rat) 304266
Molecular Function:
heterotrimeric G-protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support