Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 2
The guanylate-binding proteins (GBPs) are a family of interferon-induced GTP-binding proteins function in innate immunity against microbial and viral pathogens. In humans, there are seven GBPs from hGBP1 to hGBP7. Human GBP1 and GBP2 have been shown to exhibit antiviral activity as well as being able to regulate the inhibition of proliferation and invasion of endothelial cells in response to interferon (IFN). GBP5 promotes selective NALP3 inflammasome responses to pathogenic bacteria and soluble but not crystalline inflammasome priming agents.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: GBP-5; GBP-TA antigen; GTP-binding protein 5; Guanine nucleotide-binding protein 5; Guanylate-binding protein 5
Gene Aliases: GBP-5; GBP5; UNQ2427/PRO4987
UniProt ID: (Human) Q96PP8
Entrez Gene ID: (Human) 115362
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support