Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
The recommended shelf life for this product is 1 year from date of receipt.
This gene encodes a protein that may bind to GC-rich DNA sequences, which suggests its involvement in the regulation of transcription. Alternative splicing of this gene results in multiple transcript variants.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: functional spliceosome-associated protein 105; GC-rich sequence DNA-binding factor 1; GC-rich sequence DNA-binding factor candidate; PAX3- and PAX7-binding protein 1
Gene Aliases: BM020; C21orf66; FSAP105; GCFC; GCFC1; PAXBP1
UniProt ID: (Human) Q96DU8
Entrez Gene ID: (Human) 94104
Molecular Function:
chromatin/chromatin-binding, or -regulatory protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support