Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
Specificity: This antibody binds to the HLENEVARLKK peptide from the leucine-zipper domain of the S. cerevisiae GCN4 transcriptional activator with picomolar affinity.
GCN4 is a transcription factor protein in yeast that plays a crucial role in the regulation of genes involved in amino acid biosynthesis, such as HIS4, HIS7, HIS3, ARG1, ARG3, ARG4, and ARG5,6. The protein contains a basic leucine zipper (bZIP) domain that enables it to bind to specific DNA sequences, thereby regulating the transcription of target genes. The protein is also involved in the regulation of other cellular processes, including stress response and nutrient signaling, such as GSH1, YAP1, and TRX2. Its structure is composed of a dimeric coiled-coil domain, which allows for protein-protein interactions, and a DNA-binding bZIP domain.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Amino acid biosynthesis regulatory protein; General control protein GCN4; General control transcription factor GCN4
Gene Aliases: AAS101; AAS3; ARG9; GCN4; YEL009C
UniProt ID: (Baker's yeast) P03069
Entrez Gene ID: (Baker's yeast) 856709
Molecular Function:
basic leucine zipper transcription factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support