Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MLRTLLRRRL FSYPTKYYFM VLVLSLITFS VLRIHQKPEF VSVRHLELAG ENPSSDINCT KVLQGDVNEI QKVKLEILTV KFKKRPRWTP DDYINMTSDC SSFIKRRKYI VEPLSKEEAE FPIAYSIVVH HKIEMLDRLL RAIYMPQNFY CIHVDTKSED SYLAAVMGIA SCFSNVFVAS RLESVVYASW SRVQADLNCM KDLYAMSANW KYLINLCGMD FPIKTNLEIV RKLKLLMGEN NLETERMPSH KEERWKKRYE VVNGKLTNTG TVKMLPPLET PLFSGSAYFV VSREYVGYVL QNEKIQKLME WAQDTYSPDE YLWATIQRIP EVPGSLPASH KYDLSDMQAV ARFVKWQYFE GDVSKGAPYP PCDGVHVRSV CIFGAGDLNW MLRKHHLFAN KFDVDVDLFA IQCLDEHLRH KALETLKH
Glycosylation is one of the most universal but at the same time complex protein modifications. Modification with sugar moeties can be both co- translational and post- translational, occurring in the endoplasmatic reticulum and golgi. Three different forms of glycosylation can be distinguished: N-linked oligosaccharides, O-linked oligosaccharides and glycosyl- phosphatidylinositol (GPI-) anchors. Glycosylation results in thousands of distinct, bioactive glycoproteins resident throughout the cell that strongly determine protein-protein, carbohydrate-protein, membrane, and adhesion properties. Diseases associated with glycosylation defects include Congenital disorders of glycosylation, (CDG), also known as carbohydrate deficient glycoprotein syndromes, and diseases associated with advanced aging.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: beta-1,3-galactosyl-O-glycosyl-glycoprotein beta-1,6-N--acetylglucosaminyltransferase; Beta-1,3-galactosyl-O-glycosyl-glycoprotein beta-1,6-N-acetylglucosaminyltransferase; beta-1,6-N-acetylglucosaminyltransferase; C2GlcNAcT; C2GNT; C2GnT-L; Core 2 beta-1,6-N-acetylglucosaminyltransferase; core 2 beta-1,6-N-acetylglucosaminyltransferase I; core 2 beta1,6 N-acetylglucosaminyltransferase-I; core 2 branching enzyme; core 2 GnT; Core 2-branching enzyme; Core2-GlcNAc-transferase; glucosaminyl (N-acetyl) transferase 1, core 2 (beta-1,6-N-acetylglucosaminyltransferase); Leukocyte type core 2 beta-1,6-N-acetylglucosaminyltransferase
Gene Aliases: C2GNT; C2GNT-L; C2GNT1; G6NT; GCNT1; NACGT2; NAGCT2
UniProt ID: (Human) Q02742
Entrez Gene ID: (Human) 2650
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support