Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Peptide Sequence: ACLLTGSLFV LLRVFSFEPV PSCRALQVLK PRDRISAIAH RGGSHDAPEN TLAAIRQAAK NGATGVELDI EFTSDGIPVL MHDNTVDRTT DGTGRLCDLT FEQIRKLN
Has glycerophosphoinositol phosphodiesterase activity. Has little or no activity towards glycerophosphocholine. GDE1 activity can be modulated by G-protein signaling pathways.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Glycerophosphodiester phosphodiesterase 1; Glycerophosphoinositol glycerophosphodiesterase GDE1; Lysophospholipase D GDE1; membrane interacting protein of RGS16; Membrane-interacting protein of RGS16; PAMCI; PCIP1; RGS16-interacting membrane protein
Gene Aliases: 363E6.2; GDE1; MIR16
UniProt ID: (Human) Q9NZC3
Entrez Gene ID: (Human) 51573
Molecular Function:
phosphodiesterase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support