Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MGAVWSALLV GGGLAGALFV WLLRGGPGDT GKDGDAEQEK DAPLGGAAIP GGHQSGSSGL SPGPSGQELV TKPEHLQESN GHLISKTKDL GKLQAASWRL QNPSREVCDN SREHVPSGQF PDTEAPATSE TSNSRSYSEV SRNESLESPM GEWGFQKGQE ISAKAATCFA EKLPSSNLLK NRAKEEMSLS DLNSQDRVDH EEWEMVPRHS SWGDVGVGGS LKAPVLNLNQ GMDNGRSTLV EARGQQVHGK MERVAVMPAG SQQVSVRFQV HYVTSTDVQF IAVTGDHECL GRWNTYIPLH YNKDGFWSHS IFLPADTVVE WKFVLVENGG VTRWEECSNR FLETGHEDKV VHAWWGIH
Acts as a cargo receptor for glycogen. Delivers its cargo to an autophagic pathway called glycophagy, resulting in the transport of glycogen to lysosomes. [UniProt]
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: genethonin 1; Genethonin-1; Glycophagy cargo receptor STBD1; Starch-binding domain-containing protein 1
Gene Aliases: GENEX3414; GENX-3414; STBD1
UniProt ID: (Human) O95210
Entrez Gene ID: (Human) 8987
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support