Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Promotes matrix assembly.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Glucose-fructose oxidoreductase domain-containing protein 2
Gene Aliases: 5730466C23Rik; GFOD2; RGD1311198; UNQ9430/PRO34691
UniProt ID: (Human) Q9BQ07, (Mouse) Q9CYH5
Entrez Gene ID: (Human) 81577, (Rat) 307801, (Mouse) 70575
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support