FIGURE: 1 / 2
Immunogen sequence: RGSPLLIGVR SKYKLSTEQI PILYRTCTLE NVKNICKTRM KRLDSSACLH AVGDKAVEFF F; Positive Samples: 293T, HeLa, A375, NIH/3T3, Mouse heart, Mouse pancreas, Mouse testis, Rat heart
GFPT2 controls the flux of glucose into the hexosamine pathway. This protein most likely involved in regulating the availability of precursors for N-and O-linked glycosylation of proteins.
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: D-fructose-6-phosphate amidotransferase 2; GFAT 2; GFAT2; glucosamine--fructose-6-phosphate aminotransferase [isomerizing] 2; Glutamine--fructose-6-phosphate aminotransferase [isomerizing] 2; glutamine: fructose-6-phosphate aminotransferase 2; glutamine:fructose 6 phosphate amidotransferase 2; Glutamine:fructose-6-phosphate amidotransferase 2; Hexosephosphate aminotransferase 2
Gene Aliases: AI480523; GFAT; GFAT 2; GFAT2; GFPT2
UniProt ID: (Human) O94808, (Rat) Q4KMC4, (Mouse) Q9Z2Z9
Entrez Gene ID: (Human) 9945, (Rat) 360518, (Mouse) 14584
Molecular Function: transaminase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support