Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MARGYGATVS LVLLGLGLAL AVIVLAVVLS RHQAPCGPQA FAHAAVAADS KVCSDIGRAI LQQQGSPVDA TIAALVCTSV VNPQSMGLGG GVIFTIYNVT TGKVEVINAR ETVPASHAPS LLDQCAQALP LGTGAQWIGV PGELRGYAEA HRRHGRLPWA QLFQPTIALL RGGHVVAPVL SRFLHNSILR PSLQASTLRQ LFFNGTEPLR PQDPLPWPAL ATTLETVATE GVEVFYTGRL GQMLVEDIAK EGSQLTLQDL AKFQPEVVDA LEVPLGDYTL YSPPPPAGGA ILSFILNVLR GFNFSTESMA RPEGRVNVYH HLVETLKFAK GQRWRLGDPR SHPKLQNASR DLLGETLAQL IRQQIDGRGD HQLSHYSLAE AWGHGTGTSH VSVLGEDGSA VAATSTINTP FGAMVYSPRT GIILNNELLD LCERCPRGSG TTPSPAVSGD RVGGAPGRCW PPVPGERSPS SMVPSILINK AQGSKLVIGG AGGELIISAV AQAIMSKLWL GFDLRAAIAA PILHVNSKGC VEYEPNFSQE VQRGLQDRGQ NQTQRPFFLN VVQAVSQEGA CVYAVSDLRK SGEAAGY
This gene is a member of a gene family that encodes gamma-glutamyl transpeptidase enzymes. This enzyme consists of a heavy and a light chain, and is able to hydrolyze the gamma-glutamyl moiety of glutathione. It converts leukotriene C4 to leukotriene D4, however, it doesn't convert synthetic substrates that are commonly used to assay gamma-glutamyl transpeptidase. Three transcript variants encoding different isoforms have been found for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: gamma-glutamyl cleaving enzyme; Gamma-glutamyl transpeptidase-related enzyme; gamma-glutamyl transpeptidase-related protein; Gamma-glutamyltransferase 5; Gamma-glutamyltransferase-like activity 1; Gamma-glutamyltranspeptidase 5; GGT 5; GGT-rel; glutathione hydrolase 5; Glutathione hydrolase 5 proenzyme; Leukotriene-C4 hydrolase
Gene Aliases: GGL; GGT 5; GGT-REL; GGT5; GGTLA1
UniProt ID: (Human) P36269
Entrez Gene ID: (Human) 2687
Molecular Function:
protease
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support