Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
The antibody is specific to GHRHR.
Growth Hormone Releasing Hormone (GHRH) is a member of a family of peptide hormones that also includes VIP (vasoactive intestinal polypeptide) and PACAP (pituitary adenylate cyclase-activating polypeptide). It is a hypothalamic peptide that binds to G-protein coupled receptors to stimulate secretion and synthesis of growth hormone (GH). GHRH receptor proteins have been identified on the plasma membrane of primary human tumor cells, and are thought to be involved in the growth of a variety of cancer cells.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: GHRH receptor; Ghrh-r; GHRHRpsv; GRF receptor; GRFR; Growth hormone - releasing receptor; Growth hormone-releasing factor receptor; Growth hormone-releasing hormone receptor
Gene Aliases: GHRFR; GHRHR; GHRHREC; GRFR; IGHD1B; lit; little
UniProt ID: (Human) Q02643, (Rat) Q02644, (Mouse) P32082
Entrez Gene ID: (Human) 2692, (Rat) 25321, (Mouse) 14602
Molecular Function:
transmembrane signal receptor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support