Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
PA1-32875 detects GHSR from human samples.
Growth hormone secretagogue receptor 1a (GHSR) is a member of the G-protein coupled receptor family. GHSR may play a role in energy homeostasis and regulation of body weight. Two identified transcript variants are expressed in several tissues and are evolutionary conserved in fish and swine. One transcript, 1a, excises an intron and encodes the functional protein; this protein is the receptor for the Ghrelin ligand and defines a neuroendocrine pathway for growth hormone release. The second transcript (1b) retains the intron and does not function as a receptor for Ghrelin; however, it may function to attenuate activity of isoform 1a. Mutations to the gene can cause isolated partial growth hormone deficiency.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: GH-releasing peptide receptor; Ghrelin receptor; GHRP; Ghrp receptor; GHS receptor; GHS-R; Growth hormone secretagogue receptor type 1
Gene Aliases: GHDP; GHSR
UniProt ID: (Human) Q92847
Entrez Gene ID: (Human) 2693
Molecular Function: G-protein coupled receptor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support