Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Promotes absorption of the essential vitamin cobalamin (Cbl) in the ileum. After interaction with CUBN, the GIF-cobalamin complex is internalized via receptor-mediated endocytosis.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Cobalamin binding intrinsic factor; Gastric intrinsic factor; gastric intrinsic factor (vitamin B synthesis); IF; INF; Intrinsic factor
Gene Aliases: AV073125; CBLIF; GIF; IF; IFMH; INF; TCN3
UniProt ID: (Human) P27352, (Mouse) P52787
Entrez Gene ID: (Human) 2694, (Mouse) 14603
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support