Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Antibody detects endogenous levels of total GIMAP5.
This gene encodes a protein belonging to the GTP-binding superfamily and to the immuno-associated nucleotide (IAN) subfamily of nucleotide-binding proteins. In humans, the IAN subfamily genes are located in a cluster at 7q36.1.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: GTPase IMAP family member 5; IAN-4; IAN-5; immune associated nucleotide 4 like 1; Immune-associated nucleotide-binding protein 5; immunity associated protein 3; Immunity-associated nucleotide 4 protein; Immunity-associated nucleotide 4-like 1 protein; Immunity-associated nucleotide 5 protein; Immunity-associated protein 3; inhibitor of radiation- and OA-induced apoptosis, Irod/Ian5; lymphopenia
Gene Aliases: GIMAP5; HIMAP3; IAN-5; IAN4; IAN4L1; IAN5; Iddm1; IMAP3; IROD; Lyp
UniProt ID: (Human) Q96F15, (Rat) Q8K3L6
Entrez Gene ID: (Human) 55340, (Rat) 246774
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support